missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethoxy)acetophenone, 98%, Thermo Scientific™

Catalog Number p-4090068
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
85013-98-5
C9H7F3O2
204.15
MFCD00042404
MOEXTBIPPMLEFX-UHFFFAOYSA-N
4'-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethanone, 4-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethan-1-one, 4'-trifluoromethoxyacetophenone, 1-4-trifluoromethoxy-phenyl-ethanone, ethanone, 1-4-trifluoromethoxy phenyl, 4-trifluoromethoxyacetophenone
522821
1-[4-(trifluoromethoxy)phenyl]ethanone
CC(=O)C1=CC=C(OC(F)(F)F)C=C1
This item is not returnable. View return policy
85013-98-5
C9H7F3O2
204.15
MFCD00042404
MOEXTBIPPMLEFX-UHFFFAOYSA-N
4'-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethanone, 4-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethan-1-one, 4'-trifluoromethoxyacetophenone, 1-4-trifluoromethoxy-phenyl-ethanone, ethanone, 1-4-trifluoromethoxy phenyl, 4-trifluoromethoxyacetophenone
522821
1-[4-(trifluoromethoxy)phenyl]ethanone
CC(=O)C1=CC=C(OC(F)(F)F)C=C1
CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.2780g/mL
Boiling Point 100.0°C to 102.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4COCH3
Quantity 1 g
Specific Gravity 1.278
Formula Weight 204.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4'-(Trifluoromethoxy)acetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 4'-(Trifluoromethoxy)acetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'