missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(trans-4-n-Propylcyclohexyl)benzeneboronic acid, 98%

Brand:  Alfa Aesar™ H64464.03

Code : NEW

Additional Details : CAS Number : 146862-02-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458936

  • £20.35 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

4-(trans-4-n-Propylcyclohexyl)benzeneboronic acid
246.16
10090337
146862-02-4
MFCD06658197
4-(trans-4-Propylcyclohexyl)phenylboronic acid

Specifications

4-(trans-4-n-Propylcyclohexyl)benzeneboronic acid
Solid
246.16
10090337
1g
146862-02-4
98%
MFCD06658197
4-(trans-4-Propylcyclohexyl)phenylboronic acid
Videos
SDS