missing translation for 'onlineSavingsMsg'
Learn More

4-Tolylboronic acid, 97%, ACROS Organics™

£25.00 - £320.70

Chemical Identifiers

CAS 5720-05-8
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00039138
InChI Key BIWQNIMLAISTBV-UHFFFAOYSA-N
Synonym 4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid
PubChem CID 79799
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10536841 SDS View Documents Acros Organics
344430010
1g Glass bottle £25.00
1g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
10693712 SDS View Documents Acros Organics
344430050
5g Glass bottle £104.70
5g View alternative products
Save Up to 
»
EU Stock 3 Please to purchase this item. Need a web account? Register with us today!
10016263 SDS View Documents Acros Organics
344430250
25g Glass bottle £320.70
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 03-09-2019
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

5720-05-8
135.96
BIWQNIMLAISTBV-UHFFFAOYSA-N
79799
C7H9BO2
MFCD00039138
4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid

Specifications

4-Tolylboronic acid
Glass bottle
1g
96.0
97%
CH3C6H4B(OH)
4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid
135.96
135.96
≥96.0%
Authentic
White
5720-05-8
100.0
C7H9BO2
MFCD00039138
BIWQNIMLAISTBV-UHFFFAOYSA-N
79799
Crystalline Powder
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Tolylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.