missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Thien-2-yltetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | 4-thiophen-2-yloxane-4-carboxylic acid |
Formula Weight | 212.27 |
Percent Purity | 97% |
Chemical Name or Material | 4-thien-2-yltetrahydropyran-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'