missing translation for 'onlineSavingsMsg'
Learn More

4-(thien-2-ylmethyl)aniline, Thermo Scientific™

Catalog Number p-4391278
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
129136-65-8
C11H11NS
189.276
PIJZLXNJWCKMSA-UHFFFAOYSA-N
4-thien-2-ylmethyl aniline, 4-thiophen-2-ylmethyl aniline, 4-thiophen-2-yl methyl aniline, 4-2-thienylmethyl phenylamine
20063869
4-(thiophen-2-ylmethyl)aniline
C1=CSC(=C1)CC2=CC=C(C=C2)N
This item is not returnable. View return policy
129136-65-8
C11H11NS
189.276
PIJZLXNJWCKMSA-UHFFFAOYSA-N
4-thien-2-ylmethyl aniline, 4-thiophen-2-ylmethyl aniline, 4-thiophen-2-yl methyl aniline, 4-2-thienylmethyl phenylamine
20063869
4-(thiophen-2-ylmethyl)aniline
C1=CSC(=C1)CC2=CC=C(C=C2)N
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.28
Percent Purity 95%
Chemical Name or Material 4-(Thien-2-ylmethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'