missing translation for 'onlineSavingsMsg'
Learn More

4-(Tetrazol-5-yl)phenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 11936761
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
179942-55-3
C7H7BN4O2
189.97
MFCD06739099 MFCD11044435
DXUPJOQUAAVAGV-UHFFFAOYSA-N
4-2h-tetrazol-5-yl phenyl boronic acid, 4-2h-tetrazol-5-yl phenylboronic acid, 4-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl-phenylboronic acid, 4-1h-tetrazol-5-yl-phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenyl boronic acid, 4-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-1htetrazol-5-yl phenylboronic acid
46737995
[4-(2H-tetrazol-5-yl)phenyl]boronic acid
OB(O)C1=CC=C(C=C1)C1=NNN=N1
This item is not returnable. View return policy
179942-55-3
C7H7BN4O2
189.97
MFCD06739099 MFCD11044435
DXUPJOQUAAVAGV-UHFFFAOYSA-N
4-2h-tetrazol-5-yl phenyl boronic acid, 4-2h-tetrazol-5-yl phenylboronic acid, 4-tetrazol-5-yl phenylboronic acid, 4-2h-tetrazol-5-yl-phenylboronic acid, 4-1h-tetrazol-5-yl-phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenyl boronic acid, 4-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-1htetrazol-5-yl phenylboronic acid
46737995
[4-(2H-tetrazol-5-yl)phenyl]boronic acid
OB(O)C1=CC=C(C=C1)C1=NNN=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 189.97
Percent Purity 97%
Chemical Name or Material 4-(Tetrazol-5-yl)phenylboronic acid
compliance-icons
Product Identifier
  • 4-(Tetrazol-5-yl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'