missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Tetrahydropyranyl methanesulfonate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and bases.
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and bases.
Specifications
Quantity | 250 mg |
Formula Weight | 180.22 |
Percent Purity | 95% |
Chemical Name or Material | 4-Tetrahydropyranyl methanesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'