missing translation for 'onlineSavingsMsg'
Learn More

4-(Tetrahydropyran-4-yloxy)benzonitrile, 97%, Thermo Scientific™

Catalog Number p-3830103
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
884507-34-0
C12H13NO2
203.241
MFCD09025875
ITXQSYHCKPGZLV-UHFFFAOYSA-N
4-tetrahydropyran-4-yloxy benzonitrile, 4-oxan-4-yloxy benzonitrile, 4-tetrahydro-2h-pyran-4-yloxy benzonitrile, 4-oxan-4-yl oxy benzonitrile, 4-tetrahydro-2h-pyran-4-yl oxy benzonitrile, benzonitrile,4-tetrahydro-2h-pyran-4-yl oxy, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenecarbonitrile
24229527
4-(oxan-4-yloxy)benzonitrile
C1COCCC1OC2=CC=C(C=C2)C#N
This item is not returnable. View return policy
884507-34-0
C12H13NO2
203.241
MFCD09025875
ITXQSYHCKPGZLV-UHFFFAOYSA-N
4-tetrahydropyran-4-yloxy benzonitrile, 4-oxan-4-yloxy benzonitrile, 4-tetrahydro-2h-pyran-4-yloxy benzonitrile, 4-oxan-4-yl oxy benzonitrile, 4-tetrahydro-2h-pyran-4-yl oxy benzonitrile, benzonitrile,4-tetrahydro-2h-pyran-4-yl oxy, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenecarbonitrile
24229527
4-(oxan-4-yloxy)benzonitrile
C1COCCC1OC2=CC=C(C=C2)C#N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material 4-(Tetrahydropyran-4-yloxy)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'