missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Tetrahydropyran-4-yloxy)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 4-tetrahydro-2h-pyran-4-yl oxy benzoic acid, 4-oxan-4-yloxy benzoic acid, 4-tetrahydropyran-4-yloxy benzoic acid, 4-oxan-4-yl oxy benzoic acid, 4-tetrahydro-2h-pyran-4-yloxy benzoic acid, 4-tetrahydro-2h-pyran-4-yl oxy benzoicacid, benzoic acid, 4-tetrahydro-2h-pyran-4-yl oxy, benzoic acid,4-tetrahydro-2h-pyran-4-yl oxy, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzoic acid |
Formula Weight | 222.24 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Tetrahydropyran-4-yloxy)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'