missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-2700910
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
123324-71-0
C10H15BO2
178.04
MFCD01009697
MNJYZNVROSZZQC-UHFFFAOYSA-N
4-t-butylphenylboronic acid, 4-tert-butylbenzeneboronic acid, 4-tert-butyl phenyl boronic acid, 4-tert-butylphenyl boronic acid, 4-tert-butyl phenylboronic acid, 4-tert-butyl-phenyl boronic acid, 4-tert-butylphenyl boranediol, p-t-butylphenylboronic acid, 4-tertbutylphenylboronic acid
2734320
(4-tert-butylphenyl)boronic acid
CC(C)(C)C1=CC=C(C=C1)B(O)O
123324-71-0
C10H15BO2
178.04
MFCD01009697
MNJYZNVROSZZQC-UHFFFAOYSA-N
4-t-butylphenylboronic acid, 4-tert-butylbenzeneboronic acid, 4-tert-butyl phenyl boronic acid, 4-tert-butylphenyl boronic acid, 4-tert-butyl phenylboronic acid, 4-tert-butyl-phenyl boronic acid, 4-tert-butylphenyl boranediol, p-t-butylphenylboronic acid, 4-tertbutylphenylboronic acid
2734320
(4-tert-butylphenyl)boronic acid
CC(C)(C)C1=CC=C(C=C1)B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 191.0°C to 196.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4B(OH)2
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,thf,dmso,dmf,methanol
Formula Weight 178.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-tert-Butylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'