missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenylacetylene, 96%, Thermo Scientific Chemicals

Catalog Number p-3523330
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
772-38-3
C12H14
158.24
MFCD00190197
ZSYQVVKVKBVHIL-UHFFFAOYSA-N
1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene
2757302
1-tert-butyl-4-ethynylbenzene
CC(C)(C)C1=CC=C(C=C1)C#C
This item is not returnable. View return policy
772-38-3
C12H14
158.24
MFCD00190197
ZSYQVVKVKBVHIL-UHFFFAOYSA-N
1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene
2757302
1-tert-butyl-4-ethynylbenzene
CC(C)(C)C1=CC=C(C=C1)C#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Brown-Khaki to Green
Boiling Point 130.0°C (40.0 mmHg)
Flash Point 82.2°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH≡CC6H4C(CH3)3
Refractive Index 1.5280 to 1.5300
Quantity 5 g
Formula Weight 158.24
Percent Purity 96%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-tert-Butylphenylacetylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylphenylacetylene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'