missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(tert-Butyldimethylsiloxy)phenol, 97%

Brand:  Alfa Aesar™ H55556.03

Code : NEW

Additional Details : CAS Number : 108534-47-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15401858

  • £74.14 / 1g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

108534-47-0
224.38
KZTVHIZALLBXMO-UHFFFAOYSA-N
597388
CC(C)(C)[Si](C)(C)OC1=CC=C(O)C=C1
C12H20O2Si
MFCD06200713
4-tert-butyldimethylsiloxy phenol, 4-tert-butyldimethylsilyl oxy phenol, 4-t-butyldimethylsiloxy phenol, 4-tert-butyldimethylsilyloxy phenol, 4-tert-butyl dimethyl silyl oxyphenol, phenol, 4-t-butyldimethylsilyloxy, acmc-20amtw, hydroquinone, tbdms derivative, 4-tert-butyldimethylsilyloxyphenol
4-[tert-butyl(dimethyl)silyl]oxyphenol

Specifications

4-(tert-Butyldimethylsiloxy)phenol
60°C to 64°C
108534-47-0
MFCD06200713
KZTVHIZALLBXMO-UHFFFAOYSA-N
4-[tert-butyl(dimethyl)silyl]oxyphenol
597388
Powder
White
1g
C12H20O2Si
4-tert-butyldimethylsiloxy phenol, 4-tert-butyldimethylsilyl oxy phenol, 4-t-butyldimethylsiloxy phenol, 4-tert-butyldimethylsilyloxy phenol, 4-tert-butyl dimethyl silyl oxyphenol, phenol, 4-t-butyldimethylsilyloxy, acmc-20amtw, hydroquinone, tbdms derivative, 4-tert-butyldimethylsilyloxyphenol
CC(C)(C)[Si](C)(C)OC1=CC=C(O)C=C1
224.38
224.38
97%
Videos
SDS Documents
Certificates