missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcalix[4]arene, 99%, Thermo Scientific Chemicals

Catalog Number 10733465
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
60705-62-6
C44H56O4
648.93
MFCD00066280
NVKLTRSBZLYZHK-UHFFFAOYSA-N
4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17,
335377
5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
This item is not returnable. View return policy
60705-62-6
C44H56O4
648.93
MFCD00066280
NVKLTRSBZLYZHK-UHFFFAOYSA-N
4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17,
335377
5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 06, IV, 7858
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,benzene,toluene
Formula Weight 648.92
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 4-tert-Butylcalix[4]arene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'