missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcalix[4]arene, 99%, ACROS Organics™

Brand:  Acros Organics 290220250

Code : 29

Additional Details : CAS Number : 60705-62-6 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10668391

  • £107.80 / 25g


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

4-tert-Butylcalix[4]arene
98.5
99%
C44H56O4
MFCD00066280
60705-62-6
100.0
99%
648.92

Specifications

4-tert-Butylcalix[4]arene
98.5
Authentic
Glass bottle
White to light beige
99%
99%
648.92
06, IV, 7858
60705-62-6
100.0
Complies
> 300.0°C
Crystalline Powder or Flakes
25g
C44H56O4
MFCD00066280
Videos
SDS