missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzylamine, 97%

Product Code. 10667654 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10667654

Brand: Thermo Scientific Acros 391010050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39895-55-1 | C11H17N | 163.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39895-55-1
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.26
MDL Number MFCD00040754
InChI Key MPWSRGAWRAYBJK-UHFFFAOYSA-N
Synonym 4-tert-butylbenzylamine, 4-tert-butylphenyl methanamine, 4-t-butylbenzylamine, 1-4-tert-butylphenyl methanamine, 4-tert-butyl-benzylamine, 4-tert-butyl phenyl methanamine, 4-tert-butyl phenyl methylamine, 4t-butylbenzylamine, p-t-butylbenzylamine, 4-tertbutylbenzylamine
PubChem CID 2735655
IUPAC Name (4-tert-butylphenyl)methanamine
SMILES CC(C)(C)C1=CC=C(C=C1)CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9270g/mL
Boiling Point 235°C to 236°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NH2CH2C6H4C(CH3)
Refractive Index 1.5200 to 1.522
Quantity 5 g
Specific Gravity 0.927
Formula Weight 163.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.