missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylbenzyl chloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
4-tert-Butylbenzyl chloride has been used to control molecular weight and prevent microgel formation during polymerization reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-tert-Butylbenzyl chloride has been used to control molecular weight and prevent microgel formation during polymerization reaction.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
4-tert-Butylbenzyl chloride has been used to control molecular weight and prevent microgel formation during polymerization reaction.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
Specifications
Density | 0.945 |
Boiling Point | 101°C to 104°C (7 mmHg) |
Flash Point | 94° (202°F) |
Refractive Index | 1.522 |
Quantity | 5 g |
UN Number | UN1760 |
Beilstein | 471673 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 182.7 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'