missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-3757135
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15084-51-2
C10H13ClO2S
232.73
MFCD00007449
YEZADZMMVHWFIY-UHFFFAOYSA-N
4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride
139882
4-tert-butylbenzenesulfonyl chloride
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl
This item is not returnable. View return policy
15084-51-2
C10H13ClO2S
232.73
MFCD00007449
YEZADZMMVHWFIY-UHFFFAOYSA-N
4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride
139882
4-tert-butylbenzenesulfonyl chloride
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 79°C to 83°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4SO2Cl
Quantity 10 g
Formula Weight 232.73
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'