missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzaldehyde, 95%, Thermo Scientific Chemicals

Catalog Number p-4089554
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
939-97-9
C11H14O
162.23
MFCD00035742
OTXINXDGSUFPNU-UHFFFAOYSA-N
4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
70324
4-tert-butylbenzaldehyde
CC(C)(C)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
939-97-9
C11H14O
162.23
MFCD00035742
OTXINXDGSUFPNU-UHFFFAOYSA-N
4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
70324
4-tert-butylbenzaldehyde
CC(C)(C)C1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 249°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4CHO
Refractive Index 1.527
Quantity 50 mL
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble
Viscosity 0.004 Pas (20°C)
Formula Weight 162.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylbenzaldehyde
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'