missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylanisole, 97%, ACROS Organics™

Brand:  Acros Organics 371310250

Code : 29

Additional Details : CAS Number : 5396-38-3 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10774943

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

4-tert-Butylanisole
96.0
97%
C11H16O
164.25
5396-38-3
100.0
97%
(CH3)3CC6H4OCH3
MFCD00026309

Specifications

4-tert-Butylanisole
96.0
0.9380g/mL
Liquid
97%
C11H16O
(CH3)3CC6H4OCH3
164.25
Clear colorless to slightly yellow
25g
5396-38-3
100.0
Authentic
97%
Glass bottle
1.5020 to 1.5040
0.938
MFCD00026309
94°C
Videos
SDS