missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)piperidine hydrochloride, 90%, Maybridge

Brand:  Maybridge MO01152DA

Code : 31

Additional Details : CAS Number : 69682-13-9 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10520323

  • £107.00 / 1g


Estimated Shipment date
from Supplier 28-03-2018
Add to basket

Specifications

Chemical Identifiers

4-(Tert-butyl)piperidine hydrochloride
90.0
90%
177.72
4-(tert-Butyl)piperidine hydrochloride (IUPAC)
69682-13-9
90%
C9H19N·ClH
MFCD05865119

Specifications

4-(Tert-butyl)piperidine hydrochloride
90.0
90%
Amber Glass Bottle
MFCD05865119
1g
69682-13-9
90%
C9H19N·ClH
177.72
4-(tert-Butyl)piperidine hydrochloride (IUPAC)
Videos
SDS