missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)phenyl isothiocyanate, 97%, Thermo Scientific™

Catalog Number p-2700906
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
19241-24-8
C11H13NS
191.292
MFCD00022059
OCGNNCBNRBTUCG-UHFFFAOYSA-N
140503
1-tert-butyl-4-isothiocyanatobenzene
CC(C)(C)C1=CC=C(C=C1)N=C=S
This item is not returnable. View return policy
19241-24-8
C11H13NS
191.292
MFCD00022059
OCGNNCBNRBTUCG-UHFFFAOYSA-N
140503
1-tert-butyl-4-isothiocyanatobenzene
CC(C)(C)C1=CC=C(C=C1)N=C=S
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula (CH3)3CC6H4NCS
Quantity 1 g
Synonym 4-tert-butylphenyl isothiocyanate, 4-t-butylphenyl isothiocyanate, benzene,1-1,1-dimethylethyl-4-isothiocyanato, 4-tert-butyl phenyl isothiocyanate, benzene, 1-1,1-dimethylethyl-4-isothiocyanato, 4-tert-butylphenylisothiocyanate, zlchem 964, 4-t-butylphenylisothiocyanate, p-t-butylphenyl isothiocyanate, 4-tert-butylphenyl-isothiocyanate
Formula Weight 191.3
Percent Purity 97%
Chemical Name or Material 4-(tert-Butyl)phenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'