missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)benzylbromide, 97%, Thermo Scientific Chemicals

Catalog Number p-3757155
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18880-00-7
C11H15Br
227.14
MFCD00000180
QZNQSIHCDAGZIA-UHFFFAOYSA-N
4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide
87836
1-(bromomethyl)-4-tert-butylbenzene
CC(C)(C)C1=CC=C(C=C1)CBr
This item is not returnable. View return policy
18880-00-7
C11H15Br
227.14
MFCD00000180
QZNQSIHCDAGZIA-UHFFFAOYSA-N
4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide
87836
1-(bromomethyl)-4-tert-butylbenzene
CC(C)(C)C1=CC=C(C=C1)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 8°C to 12°C
Color Yellow
Density 1.2360g/mL
Boiling Point 90°C to 94°C (1.5 mmHg)
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4CH2Br
Refractive Index 1.5445 to 1.5465
Quantity 5 g
Specific Gravity 1.236
Formula Weight 227.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(tert-Butyl)benzylbromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-(tert-Butyl)benzylbromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'