missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butyl)benzene-1-carbothioamide, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Linear Formula | (CH3)3CC6H4CSNH2 |
Quantity | 1 g |
Formula Weight | 193.31 |
Percent Purity | 95% |
Chemical Name or Material | 4-(tert-Butyl)benzene-1-carbothioamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'