missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)-2,6-dimethylaniline, 95%, Maybridge

Brand:  Maybridge RDR03896EA

Code : 31

Additional Details : CAS Number : 42014-60-8 Weight : 0.06000kg Transport : UN number : 2810 Chem class : 1 Pack group : III

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10268172

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

4-(tert-Butyl)-2, 6-dimethylaniline
95.0
95%
177.29
42014-60-8
95%
C12H19N
MFCD00052181

Specifications

4-(tert-Butyl)-2, 6-dimethylaniline
95.0
95%
95%
177.29
42014-60-8
Amber Glass Bottle
10g
C12H19N
MFCD00052181
Videos
SDS