missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-tert-Butyl-2,6-diaminoanisole, 97%

Brand:  Alfa Aesar™ H55878.03

Code : NEW

Additional Details : CAS Number : 473269-70-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15432268

  • £18.66 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

4-tert-Butyl-2,6-diaminoanisole
473269-70-4
194.28
MFCD03701559

Specifications

4-tert-Butyl-2,6-diaminoanisole
Powder
194.28
White to orange
1g
473269-70-4
97%
MFCD03701559
111° to 113°C
Videos
SDS