missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-tert-Butyl-2,6-diaminoanisole, 97%

Brand:  Alfa Aesar™ H55878.14

Code : NEW

Additional Details : CAS Number : 473269-70-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15452268

  • £304.33 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-tert-Butyl-2,6-diaminoanisole
473269-70-4
194.28
MFCD03701559

Specifications

4-tert-Butyl-2,6-diaminoanisole
Powder
194.28
White to orange
25g
473269-70-4
97%
MFCD03701559
111° to 113°C
Videos
SDS