missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™

Catalog Number p-2700872
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
74370-93-7
C7H12N2S
156.247
MFCD00051644
CUWZBHVYLVGOAB-UHFFFAOYSA-N
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
2734202
4-tert-butyl-1,3-thiazol-2-amine
CC(C)(C)C1=CSC(=N1)N
This item is not returnable. View return policy
74370-93-7
C7H12N2S
156.247
MFCD00051644
CUWZBHVYLVGOAB-UHFFFAOYSA-N
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
2734202
4-tert-butyl-1,3-thiazol-2-amine
CC(C)(C)C1=CSC(=N1)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 156.25
Percent Purity 97%
Chemical Name or Material 4-(tert-Butyl)-1, 3-thiazol-2-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'