missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butoxycarbonyl)thiomorpholine-3-carboxylic acid, ≥97%, Thermo Scientific™

Catalog Number p-3110565
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
128453-98-5
C10H17NO4S
247.309
MFCD02683067
CTDIKDIZNAGMFK-UHFFFAOYSA-N
4-tert-butoxycarbonyl thiomorpholine-3-carboxylic acid, 4-boc-thiomorpholine-3-carboxylic acid, n-boc-3-thiomorpholinecarboxylic acid, thiomorpholine-3,4-dicarboxylic acid 4-tert-butyl ester, 4-tert-butoxy carbonyl thiomorpholine-3-carboxylic acid, 3,4-thiomorpholinedicarboxylicacid, 4-1,1-dimethylethyl ester, acmc-20dpph, acmc-1bvvh, +/--thiomorpholine-3,4-dicarboxylicacid4-tert-butylester, n-boc-3-thiomorpholinecarboxylicacid
2756831
4-[(2-methylpropan-2-yl)oxycarbonyl]thiomorpholine-3-carboxylic acid
CC(C)(C)OC(=O)N1CCSCC1C(=O)O
This item is not returnable. View return policy
128453-98-5
C10H17NO4S
247.309
MFCD02683067
CTDIKDIZNAGMFK-UHFFFAOYSA-N
4-tert-butoxycarbonyl thiomorpholine-3-carboxylic acid, 4-boc-thiomorpholine-3-carboxylic acid, n-boc-3-thiomorpholinecarboxylic acid, thiomorpholine-3,4-dicarboxylic acid 4-tert-butyl ester, 4-tert-butoxy carbonyl thiomorpholine-3-carboxylic acid, 3,4-thiomorpholinedicarboxylicacid, 4-1,1-dimethylethyl ester, acmc-20dpph, acmc-1bvvh, +/--thiomorpholine-3,4-dicarboxylicacid4-tert-butylester, n-boc-3-thiomorpholinecarboxylicacid
2756831
4-[(2-methylpropan-2-yl)oxycarbonyl]thiomorpholine-3-carboxylic acid
CC(C)(C)OC(=O)N1CCSCC1C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 247.31
Percent Purity 97%
Chemical Name or Material 4-(Tert-butoxycarbonyl)thiomorpholine-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'