missing translation for 'onlineSavingsMsg'
Learn More

[4-(tert-Butoxycarbonyl)piperazin-1-yl]acetic acid, 97%, Thermo Scientific™

Catalog Number 10142844
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
156478-71-6
C11H20N2O4
244.291
MFCD02682402
WZBHMXRBXXCEDD-UHFFFAOYSA-N
2-4-tert-butoxycarbonyl piperazin-1-yl acetic acid, 4-boc-1-piperazineacetic acid, 1-boc-4-carboxymethyl piperazine, 4-tert-butoxycarbonyl piperazin-1-yl acetic acid, 4-boc-piperazineacetic acid, 2-1-boc-piperazin-4-yl-acetic acid, boc-4-carboxymethyl piperazine, 1-piperazineacetic acid, 4-1,1-dimethylethoxy carbonyl, 2-4-tert-butoxy carbonyl piperazin-1-yl acetic acid, 4-carboxymethyl-piperazine-1-carboxylic acid tert-butyl ester
2735642
2-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]acetic acid
CC(C)(C)OC(=O)N1CCN(CC1)CC(=O)O
This item is not returnable. View return policy
156478-71-6
C11H20N2O4
244.291
MFCD02682402
WZBHMXRBXXCEDD-UHFFFAOYSA-N
2-4-tert-butoxycarbonyl piperazin-1-yl acetic acid, 4-boc-1-piperazineacetic acid, 1-boc-4-carboxymethyl piperazine, 4-tert-butoxycarbonyl piperazin-1-yl acetic acid, 4-boc-piperazineacetic acid, 2-1-boc-piperazin-4-yl-acetic acid, boc-4-carboxymethyl piperazine, 1-piperazineacetic acid, 4-1,1-dimethylethoxy carbonyl, 2-4-tert-butoxy carbonyl piperazin-1-yl acetic acid, 4-carboxymethyl-piperazine-1-carboxylic acid tert-butyl ester
2735642
2-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]acetic acid
CC(C)(C)OC(=O)N1CCN(CC1)CC(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 244.29
Percent Purity 97%
Chemical Name or Material [4-(Tert-butoxycarbonyl)piperazin-1-yl]acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'