missing translation for 'onlineSavingsMsg'
Learn More

4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2701115
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
77716-11-1
C11H16N2O4
240.259
MFCD02094544
GOLAEMFBWAIGRX-UHFFFAOYSA-N
4-tert-butoxycarbonyl amino-1-methyl-1h-pyrrole-2-carboxylic acid, 4-tert-butoxycarbonylamino-1-methyl-1h-pyrrole-2-carboxylic acid, boc-py-oh, 4-boc-amino-1-methylpyrrole-2-carboxylic acid, 4-tert-butoxycarbonyl amino-1-methylpyrrole-2-carboxylic acid, 4-boc-amino-1-methyl-1h-pyrrole-2-carboxylic acid, 4-tert-butoxycarbonylamino-1-methyl-1h-pyrrole-2 carboxylic acid, boc-4-amino-1-methylpyrrole-2-carboxylic acid
2735636
1-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]pyrrole-2-carboxylic acid
CC(C)(C)OC(=O)NC1=CN(C(=C1)C(=O)O)C
This item is not returnable. View return policy
77716-11-1
C11H16N2O4
240.259
MFCD02094544
GOLAEMFBWAIGRX-UHFFFAOYSA-N
4-tert-butoxycarbonyl amino-1-methyl-1h-pyrrole-2-carboxylic acid, 4-tert-butoxycarbonylamino-1-methyl-1h-pyrrole-2-carboxylic acid, boc-py-oh, 4-boc-amino-1-methylpyrrole-2-carboxylic acid, 4-tert-butoxycarbonyl amino-1-methylpyrrole-2-carboxylic acid, 4-boc-amino-1-methyl-1h-pyrrole-2-carboxylic acid, 4-tert-butoxycarbonylamino-1-methyl-1h-pyrrole-2 carboxylic acid, boc-4-amino-1-methylpyrrole-2-carboxylic acid
2735636
1-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]pyrrole-2-carboxylic acid
CC(C)(C)OC(=O)NC1=CN(C(=C1)C(=O)O)C
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Formula Weight 240.26
Percent Purity 97%
Quantity 250 mg
Chemical Name or Material 4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'