missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfophthalic Acid, 50% in Water Pract., ACROS Organics™

Brand:  Acros Organics 419930250

Code : 29

Additional Details : CAS Number : 89-08-7 Weight : 0.07500kg Transport : UN number : 3265 Chem class : 8 Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10761642

  • £20.80 / 25g


Estimated Shipment
19-06-2018
Add to basket


Specifications

Chemical Identifiers

4-Sulfophthalic acid
7732-18-5
C8H6O7S
MFCD00007494
pract.
46 to 54%
246.2

Specifications

4-Sulfophthalic acid
pract.
1.2900g/mL
46 to 54%
246.2
11, 406
1.4450 to 1.4490
92.0°C
25g
7732-18-5
Liquid
C8H6O7S
MFCD00007494
Glass bottle
1.29
Clear slightly brown
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Sulfophthalic acid, 50% in water

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS