missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfonic calix[6]arene hydrate, 95%, ACROS Organics™

Brand:  Acros Organics 290260010

Code : 29

Additional Details : CAS Number : 205652-55-7 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10703451

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Chelation/Complexation reagent
Specifications

Chemical Identifiers

4-Sulfonic calix[6]arene hydrate
94.0
≥94%
C42H36O24S6·xH2O
MFCD00216907
205652-55-7
100.0
95%
1117.13

Specifications

4-Sulfonic calix[6]arene hydrate
205652-55-7
100.0
≥94%
C42H36O24S6·xH2O
MFCD00216907
Glass bottle
1g
94.0
Powder
95%
1117.13
Authentic
Light beige to gray-brown
Videos
SDS