missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Sulfonamidophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2.5 g
10 g
50 g
Description
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 217°C to 219°C (decomposition) |
Quantity | 2.5 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 223.68 |
Percent Purity | 97% |
Chemical Name or Material | 4-Sulfonamidophenylhydrazine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'