missing translation for 'onlineSavingsMsg'
Learn More

4-(Pyrrolidin-1-ylmethyl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog Number p-3485603
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
650628-72-1
C12H15NO
189.258
MFCD07782421
MTBYCACQSYAYCC-UHFFFAOYSA-N
23090828
4-(pyrrolidin-1-ylmethyl)benzaldehyde
C1CCN(C1)CC2=CC=C(C=C2)C=O
This item is not returnable. View return policy
650628-72-1
C12H15NO
189.258
MFCD07782421
MTBYCACQSYAYCC-UHFFFAOYSA-N
23090828
4-(pyrrolidin-1-ylmethyl)benzaldehyde
C1CCN(C1)CC2=CC=C(C=C2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-pyrrolidin-1-ylmethyl benzaldehyde, 4-1-pyrrolidinylmethyl benzaldehyde, benzaldehyde, 4-1-pyrrolidinylmethyl, 4-pyrrolidin-1-yl methyl-benzaldehyde, 1-4-formylbenzyl pyrrolidine, 4-pyrrolizinomethyl benzaldehyde, 4-pyrrolidinylmethyl benzaldehyde, 4-pyrrolidin-1-ylmethyl-benzaldehyde, benzaldehyde,4-1-pyrrolidinylmethyl, 4-pyrrolidin-1-yl methyl benzaldehyde
Formula Weight 189.26
Percent Purity 97%
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'