missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidinecarboxaldehyde, 95%, Thermo Scientific™

Catalog Number 15219926
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2435-50-9
C5H4N2O
108.10
MFCD03789624
OKULHRWWYCFJAB-UHFFFAOYSA-N
pyrimidine-4-carboxaldehyde, 4-pyrimidinecarboxaldehyde, 4-pyrimidine carbaldehyde, 4-formylpyrimidine, 4-formyl-1,3-diazine, acmc-1cp8l, 4-pyrimidinecarboxaldehyde 7ci,8ci,9ci
11412355
pyrimidine-4-carbaldehyde
O=CC1=NC=NC=C1
This item is not returnable. View return policy
2435-50-9
C5H4N2O
108.10
MFCD03789624
OKULHRWWYCFJAB-UHFFFAOYSA-N
pyrimidine-4-carboxaldehyde, 4-pyrimidinecarboxaldehyde, 4-pyrimidine carbaldehyde, 4-formylpyrimidine, 4-formyl-1,3-diazine, acmc-1cp8l, 4-pyrimidinecarboxaldehyde 7ci,8ci,9ci
11412355
pyrimidine-4-carbaldehyde
O=CC1=NC=NC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 108.1
Percent Purity 95%
Chemical Name or Material 4-Pyrimidinecarboxaldehyde
compliance-icons
Product Identifier
  • 4-Pyrimidinecarboxaldehyde, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'