Learn More
Thermo Scientific Maybridge 4-Pyrimidin-5-ylbenzaldehyde, 1g, 95%
Brand: Thermo Scientific Maybridge CC44304DA
4-Pyrimidin-5-ylbenzaldehyde, 1g, 95%
Code : 31
Additional Details : CAS Number : 198084-12-7 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC44304DA
4-Pyrimidin-5-ylbenzaldehyde, 1g, 95%
Code : 31
Additional Details : CAS Number : 198084-12-7 Weight : 0.05100kg