Learn More
Thermo Scientific Maybridge 4-Pyrimidin-5-ylbenzaldehyde, 5g, 95%
Brand: Thermo Scientific Maybridge CC44304DE
4-Pyrimidin-5-ylbenzaldehyde, 5g, 95%
Code : 31
Additional Details : CAS Number : 198084-12-7 Weight : 0.05500kg
Brand: Thermo Scientific Maybridge CC44304DE
4-Pyrimidin-5-ylbenzaldehyde, 5g, 95%
Code : 31
Additional Details : CAS Number : 198084-12-7 Weight : 0.05500kg