missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylbenzoic acid, 97%, Thermo Scientific™

Catalog Number p-3522540
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
199678-12-1
C11H8N2O2
200.197
MFCD01318668
WNDAEOTYLPWXPN-UHFFFAOYSA-N
4-pyrimidin-2-yl-benzoic acid, 4-pyrimidin-2-yl benzoic acid, 4-2-pyrimidinyl benzoic acid, benzoic acid, 4-2-pyrimidinyl, benzoic acid, 4-2-pyrimidinyl-9ci, akos bar-2339, pubchem15861, acmc-1c8kl, 4-pyrimidin-2-yl-benzoicacid
1514324
4-pyrimidin-2-ylbenzoic acid
C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)O
This item is not returnable. View return policy
199678-12-1
C11H8N2O2
200.197
MFCD01318668
WNDAEOTYLPWXPN-UHFFFAOYSA-N
4-pyrimidin-2-yl-benzoic acid, 4-pyrimidin-2-yl benzoic acid, 4-2-pyrimidinyl benzoic acid, benzoic acid, 4-2-pyrimidinyl, benzoic acid, 4-2-pyrimidinyl-9ci, akos bar-2339, pubchem15861, acmc-1c8kl, 4-pyrimidin-2-yl-benzoicacid
1514324
4-pyrimidin-2-ylbenzoic acid
C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 200.2
Percent Purity 97%
Chemical Name or Material 4-Pyrimidin-2-ylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'