missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylbenzaldehyde, 95%, Maybridge

£89.00 - £290.00

Chemical Identifiers

CAS 77232-38-3
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.198
MDL Number MFCD08056282
InChI Key JUYLMPWKVWDXBE-UHFFFAOYSA-N
Synonym 4-pyrimidin-2-yl benzaldehyde, 4-2-pyrimidinyl benzaldehyde, 2-4-formylphenyl pyrimidine, 4-pyrimidin-2-yl-benzaldehyde, 4-2-pyrimidinyl-benzaldehyde, benzaldehyde,4-2-pyrimidinyl, benzaldehyde, 4-2-pyrimidinyl
PubChem CID 12703722
IUPAC Name 4-pyrimidin-2-ylbenzaldehyde
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10701684 SDS View Documents Maybridge
CC56204DA
1g Amber Glass Bottle £89.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 01-04-2020
Please to purchase this item. Need a web account? Register with us today!
10051924 SDS View Documents Maybridge
CC56204DE
5g Amber Glass Bottle £290.00
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 01-04-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

77232-38-3
184.198
JUYLMPWKVWDXBE-UHFFFAOYSA-N
12703722
C1=CN=C(N=C1)C2=CC=C(C=C2)C=O
C11H8N2O
MFCD08056282
4-pyrimidin-2-yl benzaldehyde, 4-2-pyrimidinyl benzaldehyde, 2-4-formylphenyl pyrimidine, 4-pyrimidin-2-yl-benzaldehyde, 4-2-pyrimidinyl-benzaldehyde, benzaldehyde,4-2-pyrimidinyl, benzaldehyde, 4-2-pyrimidinyl
4-pyrimidin-2-ylbenzaldehyde

Specifications

4-Pyrimidin-2-ylbenzaldehyde
1g
94.5
95%
MFCD08056282
JUYLMPWKVWDXBE-UHFFFAOYSA-N
4-pyrimidin-2-ylbenzaldehyde
12703722
95%
Amber Glass Bottle
77232-38-3
100.0
C11H8N2O
4-pyrimidin-2-yl benzaldehyde, 4-2-pyrimidinyl benzaldehyde, 2-4-formylphenyl pyrimidine, 4-pyrimidin-2-yl-benzaldehyde, 4-2-pyrimidinyl-benzaldehyde, benzaldehyde,4-2-pyrimidinyl, benzaldehyde, 4-2-pyrimidinyl
C1=CN=C(N=C1)C2=CC=C(C=C2)C=O
184.198
184.2