missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridinecarboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-3737289
22.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
872-85-5
C6H5NO
107.11
MFCD00006425
BGUWFUQJCDRPTL-UHFFFAOYSA-N
4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde
13389
pyridine-4-carbaldehyde
C1=CN=CC=C1C=O
872-85-5
C6H5NO
107.11
MFCD00006425
BGUWFUQJCDRPTL-UHFFFAOYSA-N
4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde
13389
pyridine-4-carbaldehyde
C1=CN=CC=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.1380g/mL
Boiling Point 77.0°C to 78.0°C (12.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)COH
Refractive Index 1.5420 to 1.5450
Quantity 25 g
Fieser 11,448
Specific Gravity 1.138
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: miscible with ethanol,acetone,toluene
Formula Weight 107.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Pyridinecarboxaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Pyridinecarboxaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'