missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Pyridineacetonitrile, ≥97%

Brand:  Alfa Aesar™ H64588.MD

Code : NEW

Additional Details : CAS Number : 13121-99-8 Weight : 0.00025kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15441869

  • £52.80 / 250mg


Estimated Shipment date
from Supplier 20-03-2018
Add to basket

Description

Description

Applications
A pyridine derivative used in the preparation of inhibitors of Acyl CoA, cholesterol acyltransferase as well as bone morphogenetic protein (BMP) signaling inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

4-Pyridineacetonitrile
118.14
UN3439
4-Pyridylacetonitrile
13121-99-8
MFCD03412601
471559

Specifications

4-Pyridineacetonitrile
Solid
118.14
UN3439
250mg
13121-99-8
≥97%
MFCD03412601
471559
4-Pyridylacetonitrile
Videos
SDS