missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(pyrid-2-yloxy)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 199.21 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Pyrid-2-yloxy)benzaldehyde |
Safety and Handling
Product Identifier
- 4-(Pyrid-2-yloxy)benzaldehyde
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 4-(Pyrid-2-yloxy)benzaldehyde
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'