missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyltetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-4391452
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
182491-21-0
C12H14O3
206.24
MFCD00085758
BWHJLSRDMNLSET-UHFFFAOYSA-N
4-phenyltetrahydro-2h-pyran-4-carboxylic acid, 4-phenyl-tetrahydro-pyran-4-carboxylic acid, 4-phenyltetrahydropyran-4-carboxylic acid, 4-phenyl-oxane-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-phenyl, 4-carboxy-4-phenyloxane, 4-carboxy-4-phenyltetrahydro-2h-pyran, 4-carboxytetrahydro-2h-pyran-4-yl benzene, 4-phenyltetrahydro-2h-pyran-4-carboxylicacid, 4-phenyl-tetrahydro-2h-pyran-4-carboxylic acid
4138553
4-phenyloxane-4-carboxylic acid
OC(=O)C1(CCOCC1)C1=CC=CC=C1
This item is not returnable. View return policy
182491-21-0
C12H14O3
206.24
MFCD00085758
BWHJLSRDMNLSET-UHFFFAOYSA-N
4-phenyltetrahydro-2h-pyran-4-carboxylic acid, 4-phenyl-tetrahydro-pyran-4-carboxylic acid, 4-phenyltetrahydropyran-4-carboxylic acid, 4-phenyl-oxane-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-phenyl, 4-carboxy-4-phenyloxane, 4-carboxy-4-phenyltetrahydro-2h-pyran, 4-carboxytetrahydro-2h-pyran-4-yl benzene, 4-phenyltetrahydro-2h-pyran-4-carboxylicacid, 4-phenyl-tetrahydro-2h-pyran-4-carboxylic acid
4138553
4-phenyloxane-4-carboxylic acid
OC(=O)C1(CCOCC1)C1=CC=CC=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 4-Phenyltetrahydropyran-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'