missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylpiperidine, 98%, Thermo Scientific Chemicals

Catalog Number p-3779446
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
771-99-3
C11H15N
161.25
MFCD00006002
UTBULQCHEUWJNV-UHFFFAOYSA-N
4-phenyl-piperidine, piperidine, 4-phenyl, 4-phenyl piperidine, chembl20969, 4-phenypiperdine, 4-phenyl piperidin, pubchem8009, 4-phenylpiperidine
69873
4-phenylpiperidine
C1CNCCC1C2=CC=CC=C2
This item is not returnable. View return policy
771-99-3
C11H15N
161.25
MFCD00006002
UTBULQCHEUWJNV-UHFFFAOYSA-N
4-phenyl-piperidine, piperidine, 4-phenyl, 4-phenyl piperidine, chembl20969, 4-phenypiperdine, 4-phenyl piperidin, pubchem8009, 4-phenylpiperidine
69873
4-phenylpiperidine
C1CNCCC1C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 57.0°C to 63.0°C
Color White to Brown
Boiling Point 255.0°C to 257.0°C (727.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 20, 291
Formula Weight 161.25
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Phenylpiperidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenylpiperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'