missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylpiperidine, 96%, Thermo Scientific Chemicals

Catalog Number p-7057060
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
771-99-3
C11H15N
161.248
MFCD00006002
UTBULQCHEUWJNV-UHFFFAOYSA-N
4-phenyl-piperidine, piperidine, 4-phenyl, 4-phenyl piperidine, chembl20969, 4-phenypiperdine, 4-phenyl piperidin, pubchem8009, 4-phenylpiperidine
69873
4-phenylpiperidine
C1CNCCC1C2=CC=CC=C2
This item is not returnable. View return policy
771-99-3
C11H15N
161.248
MFCD00006002
UTBULQCHEUWJNV-UHFFFAOYSA-N
4-phenyl-piperidine, piperidine, 4-phenyl, 4-phenyl piperidine, chembl20969, 4-phenypiperdine, 4-phenyl piperidin, pubchem8009, 4-phenylpiperidine
69873
4-phenylpiperidine
C1CNCCC1C2=CC=CC=C2

4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY
Melting Point 61°C to 64°C
Color Yellow to Brown
Quantity 1 g
Beilstein 124508
Solubility Information Soluble in water.
Formula Weight 161.24
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-Phenylpiperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'