missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Phenylnaphthalene-1-boronic acid, 98%

Brand:  Alfa Aesar™ H64277.06

Code : NEW

Additional Details : CAS Number : 372521-91-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15441399

  • £162.50 / 5g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Phenylnaphthalene-1-boronic acid
248.09
11441103
372521-91-0
MFCD16294428

Specifications

4-Phenylnaphthalene-1-boronic acid
372521-91-0
98%
MFCD16294428
5g
Solid
248.09
11441103
Videos
SDS