missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylcyclohexanone, 98+%, Thermo Scientific Chemicals

Catalog Number p-3758664
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4894-75-1
C12H14O
174.24
MFCD00001641
YKAYMASDSHFOGI-UHFFFAOYSA-N
4-phenylcyclohexanone, cyclohexanone, 4-phenyl, 4-phenyl-1-cyclohexanone, 4-phenyl cyclohexanone, 4-phenyl-cyclohexanone, 4-phenylcyclo-hexanone, acmc-1aoge, enamine_005496, 1-n-phenylpiperidin-4-one, 4-phenyl-cyclohexan-1-one
78605
4-phenylcyclohexan-1-one
C1CC(=O)CCC1C2=CC=CC=C2
This item is not returnable. View return policy
4894-75-1
C12H14O
174.24
MFCD00001641
YKAYMASDSHFOGI-UHFFFAOYSA-N
4-phenylcyclohexanone, cyclohexanone, 4-phenyl, 4-phenyl-1-cyclohexanone, 4-phenyl cyclohexanone, 4-phenyl-cyclohexanone, 4-phenylcyclo-hexanone, acmc-1aoge, enamine_005496, 1-n-phenylpiperidin-4-one, 4-phenyl-cyclohexan-1-one
78605
4-phenylcyclohexan-1-one
C1CC(=O)CCC1C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 77°C to 80°C
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H9(=O)
Quantity 10 g
Beilstein 07, III, 1466
Solubility Information Solubility in water: slightly soluble
Formula Weight 174.24
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylcyclohexanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'