missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyric acid, 99%, ACROS Organics™

Brand:  Acros Organics 130380250

Code : 29

Additional Details : CAS Number : 1821-12-1 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10614802

  • £20.50 / 25g


Estimated Shipment
02-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Phenylbutyric acid
99%
C6H5(CH2)3CO2H
99%
C10H12O2
164.2

Specifications

4-Phenylbutyric acid
165.0°C (10.0 mmHg)
49.0° to 52.0°C
1821-12-1
Glass bottle
99%
C10H12O2
164.2
>110°C
White to slightly yellow
25g
Authentic
Crystalline Powder
99%
C6H5(CH2)3CO2H
Videos
SDS