missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-phenylbutyl isothiocyanate, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Quantity | 1 g |
Formula Weight | 191.3 |
Percent Purity | 95% |
Chemical Name or Material | 4-Phenylbutyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'