missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylazophenol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.
Solubility
Soluble in acetone, ethanol, benzene and ether. Insoluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.
Solubility
Soluble in acetone, ethanol, benzene and ether. Insoluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | 152°C to 155°C |
Quantity | 5 g |
Solubility Information | Soluble in acetone,ethanol,benzene and ether. Insoluble in water. |
Formula Weight | 198.23 |
Percent Purity | 98% |
Chemical Name or Material | 4-Phenylazophenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'